Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane Dihydrobromide 98.0+%, TCI America™
SDP

Catalog No. B20541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20541G 1 g
B20545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20541G Supplier TCI America Supplier No. B20541G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 116258-17-4
Molecular Formula C12H18Br2N2
Molecular Weight (g/mol) 350.098
MDL Number MFCD01321292
InChI Key SOMPEQIPSQFVMO-AQEKLAMFSA-N
PubChem CID 14612542
IUPAC Name (1S,4S)-2-benzyl-2,5-diazabicyclo[2.2.1]heptane;dihydrobromide
SMILES C1C2CNC1CN2CC3=CC=CC=C3.Br.Br

Specifications

Color White
Quantity 1 g
Formula Weight 350.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane Dihydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.