Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S)-(-)-Verbenone, 94%

Catalog No. p-211502
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

CAS: 1196-01-6 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1196-01-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001343
InChI Key DCSCXTJOXBUFGB-JGVFFNPUSA-N
Synonym levoverbenone, --verbenone, l-verbenone, 1s---verbenone, --2-pinen-4-one, verbenone, unii-2xp0j7754u, 1s,5s-4,6,6-trimethylbicyclo 3.1.1 hept-3-en-2-one, verbenone, l, bicyclo 3.1.1 hept-3-en-2-one, 4,6,6-trimethyl-, 1s,5s
PubChem CID 92874
ChEBI CHEBI:78316
IUPAC Name (1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-en-4-one
SMILES CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C

Specifications

Color Yellow
Density 0.9700g/mL
Boiling Point 227°C to 228°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.497
Quantity 10 g
Beilstein 07, 161
Specific Gravity 0.97
Specific Rotation Condition − 220.00 (20.00°C c=3−4,CHCL3)
Specific Rotation -220
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with most common organic solvents
Formula Weight 150.22
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material (1S)-(-)-Verbenone, 94%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:
EINECSNumber 214-807-2
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.