Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ 2.0 ml Hydrating fluid for Pseudomonas
SDP

Catalog No. 10810322 Shop All Microbiologics Products
Click to view available options
Quantity:
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Inactivated Helix Elite Molecular Standards are used for every step of the molecular testing process. Quick dissolving lyophilized pellets may be reconstituted in buffer or transport medium. These are FDA listed and CE Marked as an In-Vitro Diagnostic (IVD) Medical Device.

  • Includes 10 vials containing 2 ml vial of microorganism hydrating fluid for Pseudomonas in each vial
  • USE WITH PSEUDOMONAS ONLY)

Specifications

Content And Storage 2°C to 8°C
Format EZ-CFU™ One Step
Manufacturing Origin United States
Product Type EZ-CFU
For Use With (Application) PSEUDOMONAS ONLY
Packaging Type Plastic Box
Quantity 10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.