Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(1,3-Benzothiazol-2-yl)acetonitrile, 97%, Thermo Scientific™
Click to view available options
:
Amber Glass Bottle; 10g
Specifications
Specifications
| CAS | 56278-50-3 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C9H6N2S |
| Quantity | 10g |
| Synonym | benzothiazole-2-acetonitrile, 2-benzo d thiazol-2-yl acetonitrile, 2-benzothiazoleacetonitrile, 2-1,3-benzothiazol-2-yl acetonitrile, benzothiazole-2-yl-acetonitrile, 1,3-benzothiazol-2-ylacetonitrile, benzothiazol-2-ylacetonitrile, 2-cyanomethyl-1,3-benzothiazole, 2-benzothiazol-2-ylethanenitrile, 2-cyanomethylbenzothiazole |
| InChI Key | ZMZSYUSDGRJZNT-UHFFFAOYSA-N |
| SMILES | C1=CC=C2C(=C1)N=C(S2)CC#N |
| IUPAC Name | 2-(1,3-benzothiazol-2-yl)acetonitrile |
| Molecular Weight (g/mol) | 174.221 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction