Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,1,3-Benzoxadiazol-4-amine, 97%, Thermo Scientific™

Catalog No. btb13048da
Click to view available options
Quantity:
1 g
5 g
10 g

Chemical Identifiers

CAS 767-63-5
Molecular Formula C6H5N3O
Molecular Weight (g/mol) 135.126
MDL Number MFCD00168448
InChI Key IPCMVRZVNJHUHR-UHFFFAOYSA-N
Synonym 4-aminobenzofurazan, benzo 1,2,5 oxadiazol-4-ylamine, 2,1,3-benzooxadiazol-4-ylamine, benzo c 1,2,5-oxadiazole-4-ylamine, benzofurazan-4-amine, benzofurazane-4-amine, acmc-209p5u, 2,1,3-benzoxadiazol-7-amine, 2,1,3-benzoxadiazol-4-ylamine, benzo c 1,2,5 oxadiazol-4-amine
PubChem CID 584570
IUPAC Name 2,1,3-benzoxadiazol-4-amine
SMILES C1=CC2=NON=C2C(=C1)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 135.13
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazol-4-amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.