Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, 99%

Catalog No. AAB2416136
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 550-99-2 | C14H15ClN2 | 246.738 g/mol

Naphazoline hydrochloride produces ionotropic effects via α-adrenoceptor stimulation. It is an activator of AR and Nischarin. It is used as a nasal decongestant and ocular vasoconstrictor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water 170 g/L, alcohol, DMSO, ethanol.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 550-99-2
Molecular Formula C14H15ClN2
Molecular Weight (g/mol) 246.738
MDL Number MFCD00012554
InChI Key DJDFFEBSKJCGHC-UHFFFAOYSA-N
Synonym naphazoline hydrochloride, albalon, naphazoline hcl, rhinantin, rhinoperd, stricylon, naphcon, niazol, rinofug, vasocon
PubChem CID 11079
ChEBI CHEBI:7470
IUPAC Name 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride
SMILES C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.Cl

Specifications

Melting Point 257°C to 261°C
Quantity 500 g
UN Number UN2811
Beilstein 3716843
Merck Index 14,6368
Solubility Information Soluble in water 170g/L,alcohol,DMSO,ethanol.
Formula Weight 246.74
Percent Purity 99%
Chemical Name or Material 2-(1-Naphthylmethyl)-2-imidazoline hydrochloride
Hazard Category H300
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 208-989-2
RTECSNumber NJ4375000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.