Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroacetamidine, 85%, Thermo Scientific™

Catalog No. p-7077501
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 354-37-0
Molecular Formula C2H3F3N2
Molecular Weight (g/mol) 112.06
MDL Number MFCD00041545
InChI Key NITMACBPVVUGOJ-UHFFFAOYSA-N
Synonym trifluoroacetamidine, trifluoroethanimidamide, 2,2,2-trifluoroacetamidine, ethanimidamide, 2,2,2-trifluoro, sntljqhjptadtaetp@, trifluoroacetoamidine, trifluoroacetamideine, trifluoro-acetamidine, acmc-20aje4, 2,2,2-trifluoroacetoamidine
PubChem CID 2776882
IUPAC Name 2,2,2-trifluoroethanimidamide
SMILES NC(=N)C(F)(F)F

Specifications

Boiling Point 35.0°C to 37.0°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 84% min. (HPLC)
Quantity 5 g
Formula Weight 112.05
Percent Purity 85%
Chemical Name or Material 2,2,2-Trifluoroacetamidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.