Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroacetamidine, 85%, Thermo Scientific™

Catalog No. AC454690050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454690050 5 g
AC454690250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454690050 Supplier Thermo Scientific Chemicals Supplier No. 454690050

Chemical Identifiers

CAS 354-37-0
Molecular Formula C2H3F3N2
Molecular Weight (g/mol) 112.06
MDL Number MFCD00041545
InChI Key NITMACBPVVUGOJ-UHFFFAOYSA-N
Synonym trifluoroacetamidine, trifluoroethanimidamide, 2,2,2-trifluoroacetamidine, ethanimidamide, 2,2,2-trifluoro, sntljqhjptadtaetp@, trifluoroacetoamidine, trifluoroacetamideine, trifluoro-acetamidine, acmc-20aje4, 2,2,2-trifluoroacetoamidine
PubChem CID 2776882
IUPAC Name 2,2,2-trifluoroethanimidamide
SMILES NC(=N)C(F)(F)F

Specifications

Boiling Point 35.0°C to 37.0°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 84% min. (HPLC)
Quantity 5 g
Formula Weight 112.05
Percent Purity 85%
Chemical Name or Material 2,2,2-Trifluoroacetamidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.