Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroacetophenone, 98%

Catalog No. p-7081427
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 434-45-7 | C8H5F3O | 174.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-45-7
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.122
MDL Number MFCD00000420
InChI Key KZJRKRQSDZGHEC-UHFFFAOYSA-N
Synonym trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09
PubChem CID 9905
IUPAC Name 2,2,2-trifluoro-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(F)(F)F

Specifications

Melting Point -40°C
Density 1.24
Boiling Point 165°C to 166°C
Flash Point 42°C (108°F)
Refractive Index 1.4595
Quantity 5 g
UN Number UN1224
Beilstein 1866286
Formula Weight 174.12
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroacetophenone
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 207-103-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.