Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethanol, 99+%

Catalog No. aaa1078818
Click to view available options
Quantity:
50 g
100 g
500 g
1000 g

Catalog No. AAA1078818

Supplier: Thermo Scientific Chemicals A1078818

Only null left
Add to Cart

CAS: 75-89-8 | C2H3F3O | 100.04 g/mol

Trifluoroethanol serves as a solvent and a raw material in organic chemistry and biology. TFE is a solvent of choice for hydrogen peroxide-mediated oxidations of sulfides. Trifluoroethanol acts as a protein denaturant. It is used in the manufacture of certain pharmaceuticals and drug substances. The drug fluromer, which is 2,2,2-trifluoro-1-vinyloxyethane, is the vinyl ether of trifluorethanol. It is an effective solvent for peptides and proteins, and used for NMR-based protein folding studies, and in the manufacture of nylon. As a source of the trifluoromethyl group, it is employed in several organic reactions, for example in Still-Gennari modification of Horner-Wadsworth-Emmons reaction (HWE) reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoroethanol serves as a solvent and a raw material in organic chemistry and biology. TFE is a solvent of choice for hydrogen peroxide-mediated oxidations of sulfides. Trifluoroethanol acts as a protein denaturant. It is used in the manufacture of certain pharmaceuticals and drug substances. The drug fluromer, which is 2,2,2-trifluoro-1-vinyloxyethane, is the vinyl ether of trifluorethanol. It is an effective solvent for peptides and proteins, and used for NMR-based protein folding studies, and in the manufacture of nylon. As a source of the trifluoromethyl group, it is employed in several organic reactions, for example in Still-Gennari modification of Horner-Wadsworth-Emmons reaction (HWE) reaction.

Solubility
Miscible with water, ethers, ketones, alcohols and chloroform.

Notes
Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
MDL Number MFCD00004672
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
Synonym trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
PubChem CID 6409
ChEBI CHEBI:42330
SMILES OCC(F)(F)F

Specifications

Melting Point -44°C
Density 1.391
Boiling Point 74°C to 75°C
Flash Point 29°C (84°F)
Refractive Index 1.29
Linear Formula CF3CH2OH
Quantity 50 g
UN Number UN1992
Beilstein 1733203
Solubility Information Miscible with water,ethers,ketones,alcohols and chloroform.
IUPAC Name 2,2,2-trifluoroethan-1-ol
Formula Weight 100.04
Percent Purity ≥99%
Chemical Name or Material 2,2,2-Trifluoroethanol
Show More Show Less
Hazard Category H226-H301+H331-H318-H360F-H373
Hazard Statement GHS H Statement
H301-H311-H318-H226-H332-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 200-913-6
RTECSNumber KM5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.