Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethyl methacrylate, 98%, stab. with 30-50 ppm 4-methoxyphenol

Catalog No. AAA1701214
Click to view available options
Quantity:
25 g

CAS: 352-87-4 | C6H7F3O2 | 168.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-87-4
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.12
MDL Number MFCD00013576,MFCD00274739
InChI Key QTKPMCIBUROOGY-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl methacrylate, trifluoroethyl methacrylate, methacrylic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylmethacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoroethyl ester, 2-methyl-acrylic acid 2,2,2-trifluoro-ethyl ester, 2-propenoic acid, 2-methyl-, trifluoroethyl ester, 2-methyl-2-propenoic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl m, acmc-1af8z
PubChem CID 9608
IUPAC Name 2,2,2-trifluoroethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(F)(F)F

Specifications

Density 1.181
Boiling Point 29°C to 30°C (40 mmHg)
Flash Point 25°C (77°F)
Odor Pungent
Refractive Index 1.361
Quantity 25 g
UN Number UN3272
Beilstein 4175872
Formula Weight 168.12
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroethyl methacrylate, stabilized with 30 to 50 ppm 4-methoxyphenol
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 206-525-3
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.