Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethyl trifluoromethanesulfonate, 97%

Catalog No. AC454970050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454970050 5 g
AC454970010 1 g
AC454970250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC454970050 Supplier Thermo Scientific Chemicals Supplier No. 454970050
Only null left
Add to Cart

CAS: 6226-25-1 | C3H2F6O3S | 232.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6226-25-1
Molecular Formula C3H2F6O3S
Molecular Weight (g/mol) 232.1
InChI Key RTMMSCJWQYWMNK-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyltrifluoromethanesulfonate, 2,2,2-trifluoroethyl triflate, methanesulfonic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoromethanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluorometanesulfonic acid, 2,2,2-trifluoroethyltrifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluoromethane sulfonate, trifluoroethyltriflate, trifluoroethyl triflate
PubChem CID 80366
IUPAC Name 2,2,2-trifluoroethyl trifluoromethanesulfonate
SMILES C(C(F)(F)F)OS(=O)(=O)C(F)(F)F

Specifications

Density 1.6100g/mL
Boiling Point 89.0°C to 91.0°C (740.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CF3SO3CH2CF3
Quantity 5 g
Specific Gravity 1.61
Formula Weight 232.1
Percent Purity 97%
Chemical Name or Material 2,2,2-Trifluoroethyl trifluoromethanesulfonate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.