Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trimethylthioacetamide, 97%

Catalog No. p-7031127
Click to view available options
Quantity:
1 g
10 g

CAS: 630-22-8 | C5H11NS | 117.21 g/mol

2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

Solubility
Soluble in water (10 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-22-8
Molecular Formula C5H11NS
Molecular Weight (g/mol) 117.21
MDL Number MFCD09742834
InChI Key FJZJUSOFGBXHCV-UHFFFAOYSA-N
Synonym 2,2,2-trimethylthioacetamide, 2,2-dimethylthiopropionamide, neopentanethioamide, propanethioamide, 2,2-dimethyl, thiopivalamide, 2,2-dimethyl-thiopropionamide, 2,2-dimethylthio propionamide, propanethioamide,2,2-dimethyl, 2-methyl-2-methylthiopropionamide, propionamide, 2-methyl-2-methylthio
PubChem CID 3031130
IUPAC Name 2,2-dimethylpropanethioamide
SMILES CC(C)(C)C(=S)N

Specifications

Melting Point 113°C to 117°C
Quantity 1 g
Solubility Information Soluble in water (10g/L) (25°C).
Formula Weight 117.21
Percent Purity 97%
Chemical Name or Material 2,2,2-Trimethylthioacetamide
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.