Learn More
2,2,3,3,3-Pentafluoro-1-propanol, 98%
Description
2,2,3,3,3-Pentafluoro-1-propanol is used as Pharmaceutical Intermediates. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,2,3,3,3-Pentafluoro-1-propanol is used as Pharmaceutical Intermediates. It can be used in agrochemical, pharmaceutical and dyestuff field etc.
Solubility
Soluble in water.21.9 g/L at 25°C
Notes
Incompatible with strong oxidizing agents, Acid chlorides, Acid anhydrides. Keep away from heat.
Specifications
Specifications
| Density | 1.505 |
| Boiling Point | 80°C to 81°C |
| Refractive Index | 1.288 |
| Quantity | 5 g |
| Beilstein | 1743133 |
| Solubility Information | Soluble in water.21.9g/L at 25°C |
| Formula Weight | 150.05 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,2,3,3,3-Pentafluoro-1-propanol |
Safety and Handling
| Hazard Category | H226-H302-H315-H319-H332-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P240-P243-P261-P264b-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P380-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| EINECSNumber | 207-012-7 |
| RTECSNumber | UB8800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.