Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane 98.0+%, TCI America™
SDP

Catalog No. N104125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N104125G 25 g
N10415G 5 g
N1041100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N104125G Supplier TCI America Supplier No. N104125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81190-28-5
Molecular Formula C7H5F9O
Molecular Weight (g/mol) 276.102
MDL Number MFCD00236085
InChI Key WUKHWLIEBSRTRH-UHFFFAOYSA-N
Synonym 3-Perfluorobutyl-1,2-epoxypropane
PubChem CID 2775819
IUPAC Name 2-(2,2,3,3,4,4,5,5,5-nonafluoropentyl)oxirane
SMILES C1C(O1)CC(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 g
UN Number 1993
Formula Weight 276.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.