Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoroheptyloxirane 98.0+%, TCI America™
SDP

Catalog No. T336525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T336525G 25 g
T33655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T336525G Supplier TCI America Supplier No. T336525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38565-52-5
Molecular Formula C9H5F13O
Molecular Weight (g/mol) 376.12
MDL Number MFCD00042356
InChI Key KGYUZRBIQCDOCN-UHFFFAOYNA-N
Synonym 3-Perfluorohexyl-1,2-epoxypropane
PubChem CID 170073
IUPAC Name 2-(2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptyl)oxirane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC1CO1

Specifications

Color Colorless
Boiling Point 154°C
Quantity 25 g
Formula Weight 376.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoroheptyloxirane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.