Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3-Tetrafluoropropyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. T224025G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 7383-71-3
Molecular Formula C6H6F4O2
Molecular Weight (g/mol) 186.11
MDL Number MFCD00042380
InChI Key VHJHZYSXJKREEE-UHFFFAOYSA-N
Synonym Acrylic Acid 2,2,3,3-Tetrafluoropropyl Ester
PubChem CID 81865
IUPAC Name 2,2,3,3-tetrafluoropropyl prop-2-enoate
SMILES FC(F)C(F)(F)COC(=O)C=C

Specifications

Color Colorless
Boiling Point 75°C
Quantity 25 g
UN Number 3272
Formula Weight 186.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3-Tetrafluoropropyl Acrylate (stabilized with MEHQ)
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.