Learn More
2,2,3,4,4,4-Hexafluorobutyl methacrylate, 96%, stab.
Description
It is an epoxy thinner and is mainly used as the modifier of the (meth)acrylate resin. Well-defined organic/inorganic hybrid fluorinated star polymers were synthesized via atom transfer radical polymerization (ATRP) of 2,2,3,4,4,4-hexafluorobutyl methacrylate (HFBMA) using octa(aminophenyl)silsesquioxane (OAPS) nano-cage as initiator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an epoxy thinner and is mainly used as the modifier of the (meth)acrylate resin. Well-defined organic/inorganic hybrid fluorinated star polymers were synthesized via atom transfer radical polymerization (ATRP) of 2,2,3,4,4,4-hexafluorobutyl methacrylate (HFBMA) using octa(aminophenyl)silsesquioxane (OAPS) nano-cage as initiator.
Solubility
Difficult to mix in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 1.352 |
| Boiling Point | 158°C |
| Flash Point | 56°C (132°F) |
| Refractive Index | 1.362 |
| Quantity | 25 g |
| UN Number | UN3272 |
| Beilstein | 2725177 |
| Solubility Information | Difficult to mix in water. |
| Formula Weight | 250.14 |
| Percent Purity | 96% |
| Show More |
Safety and Handling
| Hazard Category | H226 |
| Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.