Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,4,6,6-Hexamethylcyclotrisilazane 97.0+%, TCI America™
SDP

Catalog No. H090925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H090925ML 25 mL
H09095ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H090925ML Supplier TCI America Supplier No. H090925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1009-93-4
Molecular Formula C6H21N3Si3
Molecular Weight (g/mol) 219.51
MDL Number MFCD00039698
InChI Key WGGNJZRNHUJNEM-UHFFFAOYSA-N
Synonym Dimethylsilazane Cyclic Trimer
PubChem CID 66094
IUPAC Name 2,2,4,4,6,6-hexamethyl-1,3,5,2,4,6-triazatrisilinane
SMILES C[Si]1(N[Si](N[Si](N1)(C)C)(C)C)C

Specifications

Color Colorless
Boiling Point 188°C
Quantity 25 mL
UN Number 1760
Formula Weight 219.51
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2,4,4,6,6-Hexamethylcyclotrisilazane
EINECSNumber (5)-6044
RTECSNumber GZ4720000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.