Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,4,6,8,8-Heptamethylnonane 97.0+%, TCI America™
SDP

Catalog No. H0365100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0365100ML 100 mL
H036525ML 25 mL
H0365500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0365100ML Supplier TCI America Supplier No. H0365100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4390-04-9
Molecular Formula C16H34
Molecular Weight (g/mol) 226.45
MDL Number MFCD00008856
InChI Key VCLJODPNBNEBKW-UHFFFAOYNA-N
Synonym isocetane, nonane, 2,2,4,4,6,8,8-heptamethyl, cyprane, pubchem16042, acmc-2097fb, dsstox_cid_30668, dsstox_gsid_52101, ksc492g9j, 2,4,4,6,8,8-heptamethylnonane, nonane,2,4,4,6,8,8-heptamethyl
PubChem CID 20414
IUPAC Name 2,2,4,4,6,8,8-heptamethylnonane
SMILES CC(CC(C)(C)C)CC(C)(C)CC(C)(C)C

Specifications

Color Colorless
Boiling Point 247°C
Quantity 100 mL
Formula Weight 226.45
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,4,4,6,8,8-Heptamethylnonane
EINECSNumber (2)-0010
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.