Learn More
2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate
Description
Employed In pharma industry. As Coalescent for latex paints. Finds applications in Coatings, Nail care, Printing inks, Solvents for cosmetics and personal care, plasticizers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -50°C |
| Color | Colorless |
| Density | 0.950 g/mL |
| Boiling Point | 254°C to 261°C |
| Refractive Index | 1.442 |
| Quantity | 100 g |
| Solubility Information | Soluble in water 900mg/L. |
| Formula Weight | 216.32 |
| Physical Form | Liquid |
| Chemical Name or Material | 2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate |
Safety and Handling
| EINECSNumber | 246-771-9 |
| RTECSNumber | UF6000000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.