Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4-Trimethylpentane, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019005
Click to view available options
Quantity:
5 mL
10 mL
50 mL

2,2,4-Trimethylpentane, a branched alkane, having a low tendency to auto-ignite, is used as a reference in determining the octane number of gasoline.

TRUSTED_SUSTAINABILITY

Specifications

CAS 540-84-1
Vapor Pressure 41 mmHg (21°C)
Vapor Density 3.9 (vs. air)
Density 0.692 g/mL (at 25°C)
Physical Form Neat
Linear Formula (CH3)2 CHCH2C(CH3)3
MDL Number MFCD00008943
Melting Point -107°C
Boiling Point 98°C to 99°C
Synonym Isooctane
Quantity 10 mL
Formula Weight 114.23
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.