Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4-Trimethylpentane, puriss. p.a., Solstice

Catalog No. 60048292 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-292 2.5 L
60-048-291 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-292 Supplier Solstice Supplier No. 322912.5L

Puriss. p.a.,≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Density 0.692g/mL at 25°C
Flash Point −12°C (10.4°F)
Ignition Point 745°F
Linear Formula (CH3)2CHCH2C(CH3)3
Quantity 2.5 L
UN Number 1262
Beilstein 1696876
Vapor Pressure 41mmHg (21°C)
Formula Weight 114.23g/mol
Percent Purity ≥99.5% (GC)
Grade puriss. p.a.
Chemical Name or Material 2,2,4-Trimethylpentane
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P261-P273-P301 + P310-P331-P501
Precautionary Statement H225-H304-H315-H336-H410
EINECSNumber 208-759-1
RTECSNumber SA3320000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.