Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Biphenol, 99%

Catalog No. p-66905
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

CAS: 1806-29-7 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1806-29-7
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002210
InChI Key IMHDGJOMLMDPJN-UHFFFAOYSA-N
Synonym 2,2'-biphenol, 1,1'-biphenyl-2,2'-diol, 2,2'-dihydroxybiphenyl, biphenyl-2,2'-diol, o,o'-diphenol, o,o'-biphenol, o-dihydroxydiphenyl, o,o'-dihydroxybiphenyl, 2,2'-biphenyldiol, 2,2'-dihydroxydiphenyl
PubChem CID 15731
ChEBI CHEBI:28970
IUPAC Name 2-(2-hydroxyphenyl)phenol
SMILES OC1=CC=CC=C1C1=CC=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 108°C to 111°C
Color Beige to Beige-Gray
Density 0.8300g/mL
Boiling Point 315°C
Flash Point 154°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula HOC6H4C6H4OH
Quantity 100 g
Beilstein 06, 989
Specific Gravity 0.83
Solubility Information Solubility in water: insoluble
Formula Weight 186.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2,2′-Biphenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-303-0
RTECSNumber DV4200000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.