Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Biphenyldicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. D0864100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0864100G 100 g
D086425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0864100G Supplier TCI America Supplier No. D0864100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 482-05-3
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00002464
InChI Key GWZCCUDJHOGOSO-UHFFFAOYSA-N
Synonym diphenic acid, 1,1'-biphenyl-2,2'-dicarboxylic acid, 2,2'-biphenyldicarboxylic acid, 2,2'-bibenzoic acid, biphenyl-2,2'-dicarboxylic acid, 2,2'-dicarboxybiphenyl, 2,2'-diphenic acid, 2-2-carboxyphenyl benzoic acid, o,o'-diphenic acid, o,o'-bibenzoic acid
PubChem CID 10210
ChEBI CHEBI:23837
IUPAC Name 2-(2-carboxyphenyl)benzoic acid
SMILES C1=CC=C(C(=C1)C2=CC=CC=C2C(=O)O)C(=O)O

Specifications

Melting Point 233°C
Color White
Quantity 100 g
Formula Weight 242.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Biphenyldicarboxylic Acid
EINECSNumber (4)-0083
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.