Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Biquinoline, 98%

Catalog No. p-7081094
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 119-91-5 | C18H12N2 | 256.308 g/mol

2,2'-Biquinoline is used as a reagent for spectrophotometric determination of Cu and as an indicator for titration of organolithium reagents, giving yellow-green charge-transfer complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Biquinoline is used as a reagent for spectrophotometric determination of Cu and as an indicator for titration of organolithium reagents, giving yellow-green charge-transfer complexes.

Solubility
Sparingly soluble in water (0.001g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-91-5
Molecular Formula C18H12N2
Molecular Weight (g/mol) 256.308
MDL Number MFCD00006740
InChI Key WPTCSQBWLUUYDV-UHFFFAOYSA-N
Synonym 2,2'-biquinoline, cuproin, cuproine, 2,2'-diquinolyl, 2,2'-biquinolyl, 2,2'-quinolyl, 2-2-quinolyl quinoline, 2,2-biquinoline, 2,2-diquinolyl, 2,2;-biquinoline
PubChem CID 8412
IUPAC Name 2-quinolin-2-ylquinoline
SMILES C1=CC=C2C(=C1)C=CC(=N2)C3=NC4=CC=CC=C4C=C3

Specifications

Melting Point 193°C to 196°C
Quantity 1 g
Beilstein 187259
Solubility Information Sparingly soluble in water (0.001g/L).
Formula Weight 256.31
Percent Purity ≥98%
Chemical Name or Material 2,2'-Biquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-357-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.