Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(3-amino-4-hydroxylphenyl)propane 97.0+%, TCI America™
SDP

Catalog No. B442925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B442925G 25 g
B44295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B442925G Supplier TCI America Supplier No. B442925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1220-78-6
Molecular Formula C15H18N2O2
Molecular Weight (g/mol) 258.32
MDL Number MFCD00437413
InChI Key UHIDYCYNRPVZCK-UHFFFAOYSA-N
PubChem CID 223319
IUPAC Name 2-amino-4-[2-(3-amino-4-hydroxyphenyl)propan-2-yl]phenol
SMILES CC(C)(C1=CC=C(O)C(N)=C1)C1=CC=C(O)C(N)=C1

Specifications

Melting Point 254°C
Quantity 25 g
Synonym 4,4′-Isopropylidenebis(2-aminophenol)
Formula Weight 258.32
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(3-amino-4-hydroxylphenyl)propane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.