Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane 98.0+%, TCI America™
SDP

Catalog No. b14155g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14155G 5 g
B141525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14155G Supplier TCI America Supplier No. B14155G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83558-87-6
Molecular Formula C15H12F6N2O2
Molecular Weight (g/mol) 366.263
MDL Number MFCD00039147
InChI Key MSTZGVRUOMBULC-UHFFFAOYSA-N
Synonym 2,2-bis 3-amino-4-hydroxyphenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl bis 2-aminophenol, 2,2-bis 3-amino-4-hydroxyphenyl-hexafluoropropane, 2-amino-4-2-3-amino-4-hydroxyphenyl-1,1,1,3,3,3-hexafluoropropan-2-yl phenol, 4,4'-hexafluoroisopropylidene bis 2-aminophenol, phenol, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis 2-amino, 4,4'-1,1,1,3,3,3-hexafluoropropane-2,2-diyl bis 2-aminophenol, 2,2-bis 3-amino-4-hydrophenyl hexafluoropropane, 1,1-bis 3-amino-4-hydroxyphenyl-2,2,2-trifluoro-1-trifluoromethyl ethane
PubChem CID 632603
IUPAC Name 2-amino-4-[2-(3-amino-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]phenol
SMILES C1=CC(=C(C=C1C(C2=CC(=C(C=C2)O)N)(C(F)(F)F)C(F)(F)F)N)O

Specifications

Melting Point 243°C
Color White
Quantity 5 g
Formula Weight 366.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane
EINECSNumber (4)-1804
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.