Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(4-carboxyphenyl)hexafluoropropane 98.0+%, TCI America™
SDP

Catalog No. B140125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B140125G 25 g
B14015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B140125G Supplier TCI America Supplier No. B140125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1171-47-7
Molecular Formula C17H10F6O4
Molecular Weight (g/mol) 392.253
MDL Number MFCD00040931
InChI Key PHQYMDAUTAXXFZ-UHFFFAOYSA-N
Synonym 2,2-bis 4-carboxyphenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl dibenzoic acid, benzoic acid, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, 4-2-4-carboxyphenyl-1,1,1,3,3,3-hexafluoropropan-2-yl benzoic acid, 4,4'-hexafluoroisopropylidene bis benzoic acid, 4-1-4-carboxyphenyl-2,2,2-trifluoro-1-trifluoromethyl ethyl benzoic acid, acmc-1c1gj, 2,2-bis-4-carboxyphenyl hexafluoropropane
PubChem CID 633763
IUPAC Name 4-[2-(4-carboxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)C(C2=CC=C(C=C2)C(=O)O)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 275°C
Color White
Quantity 25 g
Formula Weight 392.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-carboxyphenyl)hexafluoropropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.