Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(hydroxymethyl)propionic acid, 98%

Catalog No. p-4510076
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum

CAS: 4767-03-7 | C5H10O4 | 134.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4767-03-7
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
InChI Key PTBDIHRZYDMNKB-UHFFFAOYSA-N

Specifications

Melting Point 185.0°C to 190.0°C
Color White
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula CH3C(CH2OH)2CO2H
Quantity 500 g
Beilstein 03,401
Merck Index 15,3277
Synonym 2,2-bis hydroxymethyl propionic acid, dimethylolpropionic acid, 3-hydroxy-2-hydroxymethyl-2-methylpropanoic acid, 2,2-bis hydroxymethyl propanoic acid, propanoic acid, 3-hydroxy-2-hydroxymethyl-2-methyl, 2,2-dimethylolpropionic acid, dimethylol propionic acid, 2,2-bis hydroxymethyl propanoicacid, unii-4nhi8v17mn, 4nhi8v17mn
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,slightly soluble in acetone,insoluble in benzene
SMILES CC(CO)(CO)C(=O)O
IUPAC Name 3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
PubChem CID 78501
Formula Weight 134.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2-Bis(hydroxymethyl)propionic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,
EINECSNumber 225-306-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.