Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Bromoethyl)-1,3-dioxolane 90.0+%, TCI America™
SDP

Catalog No. B1132250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1132250G 250 g
B113225G 25 g
B1132100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1132250G Supplier TCI America Supplier No. B1132250G

Chemical Identifiers

CAS 18742-02-4
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00003216
InChI Key GGZQLTVZPOGLCC-UHFFFAOYSA-N
Synonym 2-2-bromoethyl-1,3-dioxolane, 1,3-dioxolane, 2-2-bromoethyl, bromoethyl-1,3-dioxolane, acmc-1bol0, 2-2-bromoethyl-dioxolane, ksc496m8b, 2-bromoethyl-1,3-dioxolane, 2 2-bromoethyl-1,3-dioxolane, 2-2-bromoethyl-1,3-dioxolan
PubChem CID 87776
IUPAC Name 2-(2-bromoethyl)-1,3-dioxolane
SMILES BrCCC1OCCO1

Specifications

Color Yellow
Boiling Point 73°C
Quantity 250 g
Formula Weight 181.03
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromoethyl)-1,3-dioxolane
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.