Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Bromoethyl)-2-methyl-1,3-dioxolane 96.0+%, TCI America™
SDP

Catalog No. B159825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B159825G 25 g
B15985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B159825G Supplier TCI America Supplier No. B159825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37865-96-6
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00191321
InChI Key SZLPRRVCSGZARF-UHFFFAOYSA-N
PubChem CID 11805619
IUPAC Name 2-(2-bromoethyl)-2-methyl-1,3-dioxolane
SMILES CC1(OCCO1)CCBr

Specifications

Color Yellow
Boiling Point 74°C
Quantity 25 g
Formula Weight 195.06
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromoethyl)-2-methyl-1,3-dioxolane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.