Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. B56631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56631G 1 g
B5663200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56631G Supplier TCI America Supplier No. B56631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77989-15-2
Molecular Formula C21H14BrN3
Molecular Weight (g/mol) 388.268
MDL Number MFCD29047055
InChI Key MSTJGWCHJCZPEQ-UHFFFAOYSA-N
PubChem CID 12689834
IUPAC Name 2-(2-bromophenyl)-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC=CC=C3Br)C4=CC=CC=C4

Specifications

Melting Point 144°C
Color White
Quantity 1 g
Formula Weight 388.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.