Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Di(tert-butylperoxy)butane, 50% solution in aromatic free mineral spirit

Catalog No. p-102590
Click to view available options
Quantity:
10 mL
500 mL
Packaging:
Plastic bottle

CAS: 2167-23-9 | C12H26O4 | 234.336 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2167-23-9, 31807-55-3, 78-93-3
Molecular Formula C12H26O4
Molecular Weight (g/mol) 234.336
MDL Number MFCD00048243
InChI Key HQOVXPHOJANJBR-UHFFFAOYSA-N
Synonym 2,2-di tert-butylperoxy butane, 2,2-bis tert-butylperoxy butane, chaloxyd p 1293al, trigonox d, chaloxyd p 1200al, butane, 2,2-bis tert-butyldioxy, unii-v15k90hftj, v15k90hftj, 2,2-bis t-butylperoxy butane, di-tert-butyl sec-butylidene diperoxide
PubChem CID 221260
IUPAC Name 2,2-bis(tert-butylperoxy)butane
SMILES CCC(C)(OOC(C)(C)C)OOC(C)(C)C

Specifications

Color Undesignated
CAS Min % 49.0
CAS Max % 51.0
Density 0.8050g/mL
Solubility Information Solubility in water: immiscible. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons
Formula Weight 234.34
Percent Purity 49 to 51%
Packaging Plastic bottle
Specific Gravity 0.805
Viscosity 2.16 mPa.s (20°C)
Quantity 10 mL
Flash Point 43°C
Physical Form Solution
Chemical Name or Material 2, 2-Di(tert-butylperoxy)butane, 50% Solution in Aromatic Free Mineral Spirit
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Flammable liquid and vapour.
Heating may cause a fire.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
May be fatal if swallowed and enters airways.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-507-2
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.