Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2,2-Dichloro-1-methylcyclopropyl)benzene, 95%, Thermo Scientific™

Catalog No. AC176421000
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3591-42-2
Molecular Formula C10H10Cl2
Molecular Weight (g/mol) 201.09
MDL Number MFCD00012260
InChI Key NXYPRVQXSWVEOB-UHFFFAOYSA-N
Synonym 2,2-dichloro-1-methylcyclopropyl benzene, benzene, 2,2-dichloro-1-methylcyclopropyl, 1,1-dichloro-2-methyl-2-phenylcyclopropane, 2,2-dichloro-1-methyl cyclopropyl benzene, 2,2-dichloro-1-methyl-cyclopropyl benzene, acmc-20aok2, 1-phenyl-1-methyl-2,2-dichlorocyclopropane, 1-methyl-2,2-dichlorocyclopropylbenzene, 2,2-dichloro-1-methyl-cyclopropyl-benzene, 2,2-dichloro-1-methyl-1-phenyl-cyclopropane
PubChem CID 107129
IUPAC Name (2,2-dichloro-1-methylcyclopropyl)benzene
SMILES CC1(CC1(Cl)Cl)C2=CC=CC=C2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.5400 to 1.5420 (20°C, 589nm)
Linear Formula C6H5C3H2(CH3)Cl2
Quantity 100 g
Formula Weight 201.09
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material (2, 2-Dichloro-1-methylcyclopropyl)benzene, 95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.