Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diethoxyacetophenone, 96%

Catalog No. p-107226
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 6175-45-7 | C12H16O3 | 208.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6175-45-7
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00009659
InChI Key PIZHFBODNLEQBL-UHFFFAOYSA-N
Synonym 2,2-diethoxyacetophenone, ethanone, 2,2-diethoxy-1-phenyl, phenylglyoxal diethyl acetal, 2,2-diethoxy-1-phenylethan-1-one, alpha,alpha-diethoxyacetophenone, unii-w4bcz1mak3, 2,2-diethoxy acetophenone, ccris 5222, benzoylformaldehyde diethyl acetal, glyoxal, phenyl-, 2-diethyl acetal
PubChem CID 22555
IUPAC Name 2,2-diethoxy-1-phenylethanone
SMILES CCOC(OCC)C(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Salmon to Yellow
Density 1.0300g/mL
Boiling Point 131°C to 134°C (10.0 mmHg)
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4985 to 1.5005
Linear Formula C6H5COCH(OC2H5)2
Quantity 5 g
Beilstein 07, I, 361
Specific Gravity 1.03
Solubility Information Solubility in water: 0.16g/L (25°C)
Viscosity 7 mPa.s (20°C)
Formula Weight 208.26
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2, 2-Diethoxyacetophenone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
EINECSNumber 228-220-4
RTECSNumber MD3284000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.