Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diethoxypropionitrile 98.0+%, TCI America™
SDP

Catalog No. D286625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D286625ML 25 mL
D28665ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D286625ML Supplier TCI America Supplier No. D286625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56011-12-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00209573
InChI Key QPFCLGADWIWYFW-UHFFFAOYSA-N
PubChem CID 566609
IUPAC Name 2,2-diethoxypropanenitrile
SMILES CCOC(C)(C#N)OCC

Specifications

Color Colorless
Boiling Point 79°C
Quantity 25 mL
UN Number 1993
Formula Weight 143.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Diethoxypropionitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.