Learn More
2,2-Difluorocyclopropanemethanol, 97%
Description
2,2-Difluorocyclopropanemethanol is used as an intermediate in the preparation of (2,2-difluorocyclopropyl)methylamine. It is also used as a reagent in the synthesis of pyrrolopyridine derivatives, which finds application as an antiviral agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,2-Difluorocyclopropanemethanol is used as an intermediate in the preparation of (2,2-difluorocyclopropyl)methylamine. It is also used as a reagent in the synthesis of pyrrolopyridine derivatives, which finds application as an antiviral agent.
Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
Specifications
Specifications
| Boiling Point | 129°C |
| Quantity | 1 g |
| UN Number | UN1987 |
| Formula Weight | 108.09 |
| Percent Purity | 97% |
| Chemical Name or Material | 2,2-Difluorocyclopropanemethanol |
Safety and Handling
| missing translation for 'healthHazard1' | H226-H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.