Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2-Difluoroethanol, 97%

Click to view available options
Quantity:
1g
5g
359-13-7
C2H4F2O
82.05
MFCD00042248
VOGSDFLJZPNWHY-UHFFFAOYSA-N
2,2-difluoroethan-1-ol,2,2-difluoro-ethanol,ethanol, 2,2-difluoro,chf2ch2oh,2,2-difluroethanol,2,2-difluorethanol,2,2-difluoro ethanol,2,2-difluoroethylalcohol,difluoroethanol related,acmc-209ik0
136187
2,2-difluoroethanol
C(C(F)F)O
359-13-7
C2H4F2O
82.05
MFCD00042248
VOGSDFLJZPNWHY-UHFFFAOYSA-N
2,2-difluoroethan-1-ol,2,2-difluoro-ethanol,ethanol, 2,2-difluoro,chf2ch2oh,2,2-difluroethanol,2,2-difluorethanol,2,2-difluoro ethanol,2,2-difluoroethylalcohol,difluoroethanol related,acmc-209ik0
136187
2,2-difluoroethanol
C(C(F)F)O
Applications
2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

Solubility
soluble in chloroform, ethanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, acids.
Quantity 5g
Flash Point 3°C (37°F)
Melting Point -28°C
Boiling Point 95°C to 96°C
UN Number UN1987
Beilstein 1731617
Formula Weight 82.05
Percent Purity 97%
Chemical Name or Material 2,2-Difluoroethanol

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.