Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Difluoroethanol, 97%

Catalog No. p-7026277
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2220103 1 g
AAB2220106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2220103 Supplier Thermo Scientific Chemicals Supplier No. B2220103
Only null left
Add to Cart

CAS: 359-13-7 | C2H4F2O | 82.05 g/mol

2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

Solubility
soluble in chloroform, ethanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 359-13-7
Molecular Formula C2H4F2O
Molecular Weight (g/mol) 82.05
MDL Number MFCD00042248
InChI Key VOGSDFLJZPNWHY-UHFFFAOYSA-N
Synonym 2,2-difluoroethan-1-ol, 2,2-difluoro-ethanol, ethanol, 2,2-difluoro, chf2ch2oh, 2,2-difluroethanol, 2,2-difluorethanol, 2,2-difluoro ethanol, 2,2-difluoroethylalcohol, difluoroethanol related, acmc-209ik0
PubChem CID 136187
IUPAC Name 2,2-difluoroethanol
SMILES C(C(F)F)O

Specifications

Melting Point -28°C
Density 1.296
Boiling Point 95°C to 96°C
Flash Point 3°C (37°F)
Refractive Index 1.3335
Quantity 1 g
UN Number UN1987
Beilstein 1731617
Solubility Information soluble in chloroform,ethanol.
Formula Weight 82.05
Percent Purity 97%
Chemical Name or Material 2,2-Difluoroethanol
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.