Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dimethoxybiphenyl, 97%

Catalog No. p-7026992
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 4877-93-4 | C14H14O2 | 214.26 g/mol

2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

Solubility
Soluble in chloroform, methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4877-93-4
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00067073
InChI Key VGMKUVCDINAAFC-UHFFFAOYSA-N
PubChem CID 611816
IUPAC Name 1-methoxy-2-(2-methoxyphenyl)benzene
SMILES COC1=CC=CC=C1C1=CC=CC=C1OC

Specifications

Melting Point 152°C to 156°C
Quantity 5 g
Synonym 2,2'-dimethoxybiphenyl, 2,2'-dimethoxy-1,1'-biphenyl, 1,1'-biphenyl, 2,2'-dimethoxy, 1-methoxy-2-2-methoxyphenyl benzene, 2,2'-dimethoxy-biphenyl, biphenyl, 2,2'-dimethoxy, 2,2/'-dimethoxybiphenyl, acmc-1ao09, 1,1'-biphenyl,2,2'-dimethoxy, 2,2'-dimethoxy-1,1'-biphenyl #
Solubility Information Soluble in chloroform,methanol.
Formula Weight 214.26
Percent Purity 97%
Chemical Name or Material 2,2'-Dimethoxybiphenyl
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.