Learn More
2,2-Dimethyl-1,3-propanediol, 99%
Description
Used for protection of carbonyl groups as 4,4-dimethyl-1,3-dioxanes, often preferred to the more usual 1,3-dioxolanes due to easier separation and greater stability, for protection of carbonyl group of 4-bromo-2-propanone using p-toluenesulfonic acid in the presence of triethyl orthoformate. It is used in the synthesis of polyesters, paints, lubricants, and plasticizers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used for protection of carbonyl groups as 4,4-dimethyl-1,3-dioxanes, often preferred to the more usual 1,3-dioxolanes due to easier separation and greater stability, for protection of carbonyl group of 4-bromo-2-propanone using p-toluenesulfonic acid in the presence of triethyl orthoformate. It is used in the synthesis of polyesters, paints, lubricants, and plasticizers.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents, water/moisture.

Specifications
Specifications
Melting Point | 125°C to 130°C |
Density | 1.042 |
Boiling Point | 210°C to 212°C |
Flash Point | 103°C (217°F) |
Odor | Mild |
Quantity | 2500 g |
Beilstein | 605291 |
Sensitivity | Hygroscopic |
Merck Index | 14,6458 |
Solubility Information | Soluble in water. |
Show More |
Safety and Handling
Hazard Category | H318-H500 |
Hazard Statement | GHS H Statement H318 Causes serious eye damage. |
Precautionary Statement | P280-P305+P351+P338-P310 |
EINECSNumber | 204-781-0 |
RTECSNumber | TY5775000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.