Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine, 97%

Catalog No. AAH6105803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6105803 1 g
AAH61058MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6105803 Supplier Thermo Scientific Chemicals Supplier No. H6105803
Add to Cart

CAS: 866089-28-3 | C10H13NO | 163.22 g/mol

It possess various biological activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It possess various biological activities.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866089-28-3
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD11603419
InChI Key WILBVUMFTFLAHJ-UHFFFAOYSA-N
Synonym 2,2-dimethyl-3,4-dihydro-2h-1,4-benzoxazine, 2,2-dimethyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-2,2-dimethyl, 2,2-dimethyl-3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-2,2-dimethyl
PubChem CID 23587101
IUPAC Name 2,2-dimethyl-3,4-dihydro-1,4-benzoxazine
SMILES CC1(CNC2=CC=CC=C2O1)C

Specifications

Quantity 1 g
Formula Weight 163.22
Percent Purity 97%
Chemical Name or Material 2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.