Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethyl-4-pentenal, 97%, stabilized

Catalog No. ac295040050
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle

CAS: 5497-67-6 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5497-67-6
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00010699
InChI Key DXSDIWHOOOBQTJ-UHFFFAOYSA-N
Synonym 2,2-dimethyl-4-pentenal, 4-pentenal, 2,2-dimethyl, 2,2-dimethyl-pent-4-enal, 4-pentenal,2,2-dimethyl, zlchem 644, acmc-20aojn, pubchem22726, 2,2-dimethyl4-pentenal, 4-pentenal,2-dimethyl, 2,2-dimethy-4-pentenal
PubChem CID 79636
IUPAC Name 2,2-dimethylpent-4-enal
SMILES CC(C)(CC=C)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8250g/mL
Boiling Point 124.0°C to 125.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2C(CH3)2CHO
Refractive Index 1.4200 to 1.4220
Quantity 5 mL
Specific Gravity 0.825
Stabilizer 0.1% hydroquinone
Formula Weight 112.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 2-Dimethyl-4-pentenal
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-834-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.