Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylbutane, ≥98.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187637
Click to view available options
Quantity:
5 mL
10 mL
Featured Industries: Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 797°F
CAS 75-83-2
Vapor Pressure 5.35 psi (20°C)
Vapor Density 2.97 (vs. air)
Density 0.649 g/mL (at 25°C)
Flash Point -29°C
Physical Form Neat
Linear Formula CH3 CH2 C(CH3)3
MDL Number MFCD00009321
Melting Point -100°C
Boiling Point 50°C
Synonym Neohexane
Type Analytical Standard
Quantity 5 mL
Formula Weight 86.18
Percent Purity ≥98.0%
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.