Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylsuccinic Anhydride 98.0+%, TCI America™
SDP

Catalog No. D441725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D441725G 25 g
D44175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D441725G Supplier TCI America Supplier No. D441725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17347-61-4
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00022608
InChI Key ACJPFLIEHGFXGP-UHFFFAOYSA-N
Synonym 2,2-dimethylsuccinic anhydride, 3,3-dimethyldihydrofuran-2,5-dione, unii-tl903a7h7v, 2,5-furandione, dihydro-3,3-dimethyl, 2,2-dimethyl succinic anhydride, 2,2-dimethylsuccinicanhydride, dihydro-3,3-dimethylfuran-2,5-dione, 2,2-dimethylsuccinic acid anhydride, dihydro-3,3-dimethyl-2,5-furandione, ksc490q1j
PubChem CID 87067
IUPAC Name 3,3-dimethyloxolane-2,5-dione
SMILES CC1(C)CC(=O)OC1=O

Specifications

Melting Point 30°C
Color White
Quantity 25 g
Physical Form Crystal-Lump at 20°C
Boiling Point 220°C
Formula Weight 128.13
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2,2-Dimethylsuccinic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.