Learn More
2,2-Diphenyl-1-picrylhydrazyl (free radical), 95%
Description
2,2-Diphenyl-1-picrylhydrazyl is used as a sensitive colorimetric free radical scavenger. It serves as a general antioxidant detector. It is utilized to monitor chemical reactions involving radicals as well as a standard of the position and intensity of electron paramagnetic resonance signals. Further, it is employed in the quantitative determination of aliphatic and aromatic thiols by indirect spectroscopy and for photometric determination of tocopherol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 1 g |
Merck Index | 14,3330 |
Solubility Information | Soluble in dimethyl formamide,ethanol,chloroform,ether,carbon disulfinde,acetone,oils,carbon tetrachloride,glacial acetic acid and toluene. Insoluble in water. |
Formula Weight | 394.32 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | 2, 2-Diphenyl-1-picrylhydrazyl (Free Radical) |
Safety and Handling
Hazard Category | H317-H334 |
Hazard Statement | GHS H Statement H334-H317 |
Precautionary Statement | P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c |
EINECSNumber | 217-591-8 |
RTECSNumber | MW3250000 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.