Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dipyridyl, 99+%

Catalog No. p-4258985
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 366-18-7 | C10H8N2 | 156.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
Synonym 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
PubChem CID 1474
ChEBI CHEBI:30351
IUPAC Name 2-pyridin-2-ylpyridine
SMILES C1=CC=C(N=C1)C1=CC=CC=N1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 69.0°C to 72.0°C
Color White
Boiling Point 272.0°C to 273.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 23,199
Fieser 16,25
Merck Index 15,1241
Solubility Information Solubility in water: slightly soluble (5.5g/L 22°C). Other solubilities: very soluble in ethanol (500g/L 25°C), ether,, benzene, chloroform and petroleum ether
Viscosity 1.51 mm2/s (90°C)
Formula Weight 156.19
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2'-Dipyridyl
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or d
EINECSNumber 206-674-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.