Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dipyrrolylmethane 97.0+%, TCI America™
SDP

Catalog No. D38511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38511G 1 g
D38515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38511G Supplier TCI America Supplier No. D38511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21211-65-4
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD03931560
InChI Key PBTPREHATAFBEN-UHFFFAOYSA-N
PubChem CID 140814
ChEBI CHEBI:36319
IUPAC Name 2-[(1H-pyrrol-2-yl)methyl]-1H-pyrrole
SMILES C(C1=CC=CN1)C1=CC=CN1

Specifications

Melting Point 74°C
Color White-Yellow
Boiling Point 120°C
Quantity 1 g
Formula Weight 146.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dipyrrolylmethane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.