Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2(2-Ethoxyethoxy)ethanol, 98+%

Catalog No. AC117890250
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117890250 25 L Metal drum
AC117890010 1 L Glass Bottle
AC117890025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC117890250 Supplier Thermo Scientific Chemicals Supplier No. 117890250
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 111-90-0 | C6H14O3 | 134.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-90-0
MDL Number MFCD00002872
InChI Key XXJWXESWEXIICW-UHFFFAOYSA-N
Synonym diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
PubChem CID 8146
ChEBI CHEBI:40572
IUPAC Name 2-(2-ethoxyethoxy)ethanol
SMILES CCOCCOCCO

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -80.0°C
Color Colorless
Density 0.9900g/mL
Boiling Point 197.0°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Metal drum
Molecular Formula C6H14O3
Refractive Index 1.4260 to 1.4280
Linear Formula C2H5OCH2CH2OCH2CH2OH
Quantity 25 L
Beilstein 01, II, 520
Specific Gravity 0.99
Merck Index 15, 3134
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetone,benzene,chloroform,,ethanol,ether,pyridine,etc
Molecular Weight (g/mol) 134.17
Viscosity 3.9 mPa.s (25°C)
Formula Weight 134.17
Percent Purity 98+%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2(2-Ethoxyethoxy)ethanol
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-919-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.